2-Methyl-5-nitrobenzenesulphonamide
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Aminosulphonyl)-4-methylnitrobenzene, 4-Nitro-2-sulphamoyltoluene
OR0636-100G OR0636-10G OR0636-25G
APOSOR0636-100GEA 85.7 EUR
APOSOR0636-100G APOSOR0636-10G APOSOR0636-25G
2-Methyl-5-nitrobenzenesulphonamide
2-Methyl-5-nitrobenzenesulphonamide
Formula: C₇H₈N₂O₄S
MW: 216.214 g/mol
Melting Pt: 197…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00115503
CAS Number: 6269-91-6