N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide
Supplier: Apollo Scientific
Warning
OR0754-10MG OR0754-50MG
APOSOR0754-10MGEA 115 EUR
APOSOR0754-10MG APOSOR0754-50MG
N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide
N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide
Formula: C₁₅H₁₅N₅O
MW: 281.312 g/mol
Melting Pt: 200…201 °C
Storage Temperature: Ambient
MDL Number: MFCD01567008
CAS Number: 108825-65-6