2-[2-(Piperazin-1-yl)-ethyl]-1,3-dioxolan 98+%
Supplier: Apollo Scientific
Warning
OR0945-2G OR0945-10G
APOSOR0945-2GEA 87 EUR
APOSOR0945-2G APOSOR0945-10G
2-[2-(Piperazin-1-yl)-ethyl]-1,3-dioxolan 98+%
2-[2-(Piperazin-1-yl)-ethyl]-1,3-dioxolan
Formula: C₉H₁₈N₂O₂
MW: 186.251 g/mol
Boiling Pt: 86…96 °C (0.01 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03412119
CAS Number: 25553-77-9