5-Methyl-2,1,3-benzothiadiazole-4-sulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 4-(Chlorosulphonyl)-5-methyl-2,1,3-benzothiadiazole
OR102828-1G OR102828-5G
APOSOR102828-1GEA 143 EUR
APOSOR102828-1G APOSOR102828-5G
5-Methyl-2,1,3-benzothiadiazole-4-sulphonyl chloride
5-Methyl-2,1,3-benzothiadiazole-4-sulphonyl chloride
Formula: C₇H₅ClN₂O₂S₂
MW: 248.71 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD02232191
CAS Number: 3662-82-6
UN: 1759
ADR: 8 III