4-Aminobenzeneboronic acid, pinacol ester hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaboralan-2-yl)aniline hydrochloride
OR10341-1G OR10341-5G
APOSOR10341-1GEA 116 EUR
APOSOR10341-1G APOSOR10341-5G
4-Aminobenzeneboronic acid, pinacol ester hydrochloride
4-Aminobenzeneboronic acid, pinacol ester hydrochloride
Formula: C₁₂H₁₈BNO₂·ClH
MW: 255.549 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD02258965
CAS Number: 616227-14-6