4-(Piperazin-1-yl)thieno[2,3-d]pyrimidine
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Thieno[2,3-d]pyrimidin-4-yl)piperazine
OR103674-250MG OR103674-1G
APOSOR103674-250MGEA 109 EUR
APOSOR103674-250MG APOSOR103674-1G
4-(Piperazin-1-yl)thieno[2,3-d]pyrimidine
4-(Piperazin-1-yl)thieno[2,3-d]pyrimidine
Formula: C₁₀H₁₂N₄S
MW: 220.294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00728969
CAS Number: 373356-48-0