Ethyl 3-oxo-4-(triphenylphosphoranylidene)butanoate
Supplier: Apollo Scientific
Warning
OR10650-5G OR10650-1G
APOSOR10650-5GEA 98.6 EUR
APOSOR10650-5G APOSOR10650-1G
Ethyl 3-oxo-4-(triphenylphosphoranylidene)butanoate
Ethyl 3-oxo-4-(triphenylphosphoranylidene)butanoate
Formula: C₂₄H₂₃O₃P
Melting Pt: 101…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00192162
CAS Number: 13148-05-5