4-Hydroxy-7-methylthieno[3,2-d]pyrimidine
Supplier: Apollo Scientific
Warning
Synonyms: 7-Methylthieno[3,2-d]pyrimidin-4-ol
OR110039-1G OR110039-5G
APOSOR110039-1GEA 29 EUR
APOSOR110039-1G APOSOR110039-5G
4-Hydroxy-7-methylthieno[3,2-d]pyrimidine
4-Hydroxy-7-methylthieno[3,2-d]pyrimidine
Formula: C₇H₆N₂OS
MW: 166.2 g/mol
Melting Pt: 240…242 °C
Storage Temperature: Ambient
MDL Number: MFCD13181517
CAS Number: 175137-13-0