5-(1,3-Thiazol-2-yl)-1,3-oxazole
Supplier: Apollo Scientific
OR110065-1G OR110065-5G
APOSOR110065-1GEA 0 EUR
APOSOR110065-1G APOSOR110065-5G
5-(1,3-Thiazol-2-yl)-1,3-oxazole
5-(1,3-Thiazol-2-yl)-1,3-oxazole
Formula: C₆H₄N₂OS
MW: 152.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24850055
CAS Number: 681135-47-7