3-[(Dimethylamino)methylene]tetrahydro-4H-pyran-4-one
Supplier: Apollo Scientific
OR110074-1G
OR110074-5G
APOSOR110074-1GEA
272
EUR
APOSOR110074-1G
APOSOR110074-5G
3-[(Dimethylamino)methylene]tetrahydro-4H-pyran-4-one
3-[(Dimethylamino)methylene]tetrahydro-4H-pyran-4-one
Formula:
C₈H₁₃NO₂ MW: 155.194 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18830261 CAS Number: 727382-75-4 |