4-(1-Methyl-1H-indazol-3-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Supplier: Apollo Scientific
OR110107-1G OR110107-250MG
APOSOR110107-1GEA 348 EUR
APOSOR110107-1G APOSOR110107-250MG
4-(1-Methyl-1H-indazol-3-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one
4-(1-Methyl-1H-indazol-3-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Formula: C₁₀H₉N₅O
MW: 215.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381088
CAS Number: 1440535-43-2