6-Bromoimidazo[1,2-a]pyridine-3-carbohydrazide
Supplier: Apollo Scientific
Warning
OR110113-1G OR110113-250MG
APOSOR110113-1GEA 696 EUR
APOSOR110113-1G APOSOR110113-250MG
6-Bromoimidazo[1,2-a]pyridine-3-carbohydrazide
6-Bromoimidazo[1,2-a]pyridine-3-carbohydrazide
Formula: C₈H₇BrN₄O
MW: 255.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24850059
CAS Number: 474709-32-5