3-Bromo-N'-hydroxyimidazo[1,2-a]pyridine-6-carboximidamide
Supplier: Apollo Scientific
OR110138-1G OR110138-250MG
APOSOR110138-1GEA 290 EUR
APOSOR110138-1G APOSOR110138-250MG
3-Bromo-N'-hydroxyimidazo[1,2-a]pyridine-6-carboximidamide
3-Bromo-N'-hydroxyimidazo[1,2-a]pyridine-6-carboximidamide
Formula: C₈H₇BrN₄O
MW: 255.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20730925
CAS Number: 1227957-37-0