2-Ethyl-5-(4-nitrophenyl)-1,3,4-oxadiazole
Supplier: Apollo Scientific
OR110336-1G OR110336-250MG
APOSOR110336-1GEA 190 EUR
APOSOR110336-1G APOSOR110336-250MG
2-Ethyl-5-(4-nitrophenyl)-1,3,4-oxadiazole
2-Ethyl-5-(4-nitrophenyl)-1,3,4-oxadiazole
Formula: C₁₀H₉N₃O₃
MW: 219.197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11094977
CAS Number: 46713-46-6