Ethyl 3-methyl-2-nitrobenzoate
Supplier: Apollo Scientific
OR110352-10G OR110352-1G OR110352-5G
APOSOR110352-10GEA 0 EUR
APOSOR110352-10G APOSOR110352-1G APOSOR110352-5G
Ethyl 3-methyl-2-nitrobenzoate
Ethyl 3-methyl-2-nitrobenzoate
Formula: C₁₀H₁₁NO₄
MW: 209.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111006
CAS Number: 54064-39-0