2-Methyl-1H-imidazole-4,5-dicarbonitrile
Supplier: Apollo Scientific
Danger
OR110367-1G OR110367-250MG
APOSOR110367-1GEA 125 EUR
APOSOR110367-1G APOSOR110367-250MG
2-Methyl-1H-imidazole-4,5-dicarbonitrile
2-Methyl-1H-imidazole-4,5-dicarbonitrile
Formula: C₆H₄N₄
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD05975058
CAS Number: 40056-53-9