2-Methyl-7-nitro-2H-1,4-benzoxazin-3(4H)-one
Supplier: Apollo Scientific
OR110369-1G OR110369-5G
APOSOR110369-1GEA 58 EUR
APOSOR110369-1G APOSOR110369-5G
2-Methyl-7-nitro-2H-1,4-benzoxazin-3(4H)-one
2-Methyl-7-nitro-2H-1,4-benzoxazin-3(4H)-one
Formula: C₉H₈N₂O₄
MW: 208.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10040175
CAS Number: 85160-82-3