5,6-Dichloro-2-(chloromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
OR110384-1G OR110384-5G OR110384-10G
APOSOR110384-1GEA 149 EUR
APOSOR110384-1G APOSOR110384-5G APOSOR110384-10G
5,6-Dichloro-2-(chloromethyl)-1H-benzimidazole
5,6-Dichloro-2-(chloromethyl)-1H-benzimidazole
Formula: C₈H₅Cl₃N₂
MW: 235.498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01658453
CAS Number: 39811-03-5