3-Bromo-5-methoxy-1H-indazole
Supplier: Apollo Scientific
Warning
OR110396-1G OR110396-250MG
APOSOR110396-1GEA 224 EUR
APOSOR110396-1G APOSOR110396-250MG
3-Bromo-5-methoxy-1H-indazole
3-Bromo-5-methoxy-1H-indazole
Formula: C₈H₇BrN₂O
MW: 227.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781660
CAS Number: 885519-30-2