2-(4-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole
Supplier: Apollo Scientific
OR110416-1G OR110416-5G
APOSOR110416-1GEA 136 EUR
APOSOR110416-1G APOSOR110416-5G
2-(4-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole
2-(4-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole
Formula: C₉H₆BrClN₂O
MW: 273.514 g/mol
Melting Pt: 187…188 °C
Storage Temperature: Ambient
MDL Number: MFCD03273472
CAS Number: 568544-04-7