2-[(4-Chlorophenyl)thio]pyridin-3-amine

Supplier: Apollo Scientific

OR110468-1G OR110468-5G
APOSOR110468-1GEA 248 EUR
APOSOR110468-1G APOSOR110468-5G
2-[(4-Chlorophenyl)thio]pyridin-3-amine
2-[(4-Chlorophenyl)thio]pyridin-3-amine
Formula: C₁₁H₉ClN₂S
MW: 236.721 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01927959
CAS Number: 353277-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR