4-(4-Methylphenoxy)benzaldehyde
Supplier: Apollo Scientific
Synonyms: 4-(p-Tolyloxy)benzaldehyde
OR110516-1G OR110516-5G
APOSOR110516-1GEA 186 EUR
APOSOR110516-1G APOSOR110516-5G
4-(4-Methylphenoxy)benzaldehyde
4-(4-Methylphenoxy)benzaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06496533
CAS Number: 61343-83-7