2-Bromo-1-(2,2-diethoxyethoxy)-4-methylbenzene
Supplier: Apollo Scientific
OR110556-1G OR110556-5G
APOSOR110556-1GEA 198 EUR
APOSOR110556-1G APOSOR110556-5G
2-Bromo-1-(2,2-diethoxyethoxy)-4-methylbenzene
2-Bromo-1-(2,2-diethoxyethoxy)-4-methylbenzene
Formula: C₁₃H₁₉BrO₃
MW: 303.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19507743
CAS Number: 1379658-80-6