Methyl 3-(3-formyl-1H-indol-1-yl)propanoate
Supplier: Apollo Scientific
Warning
OR110562-1G OR110562-5G OR110562-10G
APOSOR110562-1GEA 173 EUR
APOSOR110562-1G APOSOR110562-5G APOSOR110562-10G
Methyl 3-(3-formyl-1H-indol-1-yl)propanoate
Methyl 3-(3-formyl-1H-indol-1-yl)propanoate
Formula: C₁₃H₁₃NO₃
MW: 231.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111410
CAS Number: 1227954-70-2