tert-Butyl 4-[2-(3-bromophenoxy)ethyl]piperazine-1-carboxylate
Supplier: Apollo Scientific
|
Warning
|
OR110584-1G
OR110584-5G
APOSOR110584-1GEA
0
EUR
APOSOR110584-1G
APOSOR110584-5G
tert-Butyl 4-[2-(3-bromophenoxy)ethyl]piperazine-1-carboxylate
tert-Butyl 4-[2-(3-bromophenoxy)ethyl]piperazine-1-carboxylate
Formula:
C₁₇H₂₅BrN₂O₃ MW: 385.296 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11973601 CAS Number: 1227954-64-4 |