4-(4-Bromo-1H-pyrazol-1-yl)aniline
Supplier: Apollo Scientific
OR110601-1G OR110601-5G
APOSOR110601-1GEA 149 EUR
APOSOR110601-1G APOSOR110601-5G
4-(4-Bromo-1H-pyrazol-1-yl)aniline
4-(4-Bromo-1H-pyrazol-1-yl)aniline
Formula: C₉H₈BrN₃
MW: 238.084 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436686
CAS Number: 681441-17-8