2,2-Dibromo-1-(4-morpholin-4-ylphenyl)ethanone
Supplier: Apollo Scientific
OR110630-1G OR110630-10G OR110630-5G
APOSOR110630-1GEA 198 EUR
APOSOR110630-1G APOSOR110630-10G APOSOR110630-5G
2,2-Dibromo-1-(4-morpholin-4-ylphenyl)ethanone
2,2-Dibromo-1-(4-morpholin-4-ylphenyl)ethanone
Formula: C₁₂H₁₃Br₂NO₂
MW: 363.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059475
CAS Number: 210832-82-9