4,5-Dibromothiophene-2-sulphonamide
Supplier: Apollo Scientific
OR110673-1G OR110673-5G
APOSOR110673-1GEA 232 EUR
APOSOR110673-1G APOSOR110673-5G
4,5-Dibromothiophene-2-sulphonamide
4,5-Dibromothiophene-2-sulphonamide
Formula: C₄H₃Br₂NO₂S₂
MW: 321.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00208579
CAS Number: 77893-69-7