N'-Hydroxy-4-propoxybenzenecarboximidamide
Supplier: Apollo Scientific
OR110686-1G OR110686-5G
APOSOR110686-1GEA 112 EUR
APOSOR110686-1G APOSOR110686-5G
N'-Hydroxy-4-propoxybenzenecarboximidamide
N'-Hydroxy-4-propoxybenzenecarboximidamide
Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664423
CAS Number: 145259-49-0