2,2-Dimethyl-7-nitro-2H-chromen-6-amine
Supplier: Apollo Scientific
Warning
OR110836-1G OR110836-250MG OR110836-5G
APOSOR110836-1GEA 24.8 EUR
APOSOR110836-1G APOSOR110836-250MG APOSOR110836-5G
2,2-Dimethyl-7-nitro-2H-chromen-6-amine
2,2-Dimethyl-7-nitro-2H-chromen-6-amine
Formula: C₁₁H₁₂N₂O₃
MW: 220.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619604
CAS Number: 64169-75-1