2,2,2-Trichloro-1-(6-chloroimidazo[1,2-a]pyridin-3-yl)ethanone
Supplier: Apollo Scientific
Warning
OR110890-1G OR110890-5G OR110890-10G
APOSOR110890-1GEA 186 EUR
APOSOR110890-1G APOSOR110890-5G APOSOR110890-10G
2,2,2-Trichloro-1-(6-chloroimidazo[1,2-a]pyridin-3-yl)ethanone
2,2,2-Trichloro-1-(6-chloroimidazo[1,2-a]pyridin-3-yl)ethanone
Formula: C₉H₄Cl₄N₂O
MW: 297.953 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25976397
CAS Number: 1823257-78-8