4-Chloro-N-(2-methyl-3-nitrophenyl)butanamide
Supplier: Apollo Scientific
OR110900-10G OR110900-1G OR110900-5G
APOSOR110900-10GEA 0 EUR
APOSOR110900-10G APOSOR110900-1G APOSOR110900-5G
4-Chloro-N-(2-methyl-3-nitrophenyl)butanamide
4-Chloro-N-(2-methyl-3-nitrophenyl)butanamide
Formula: C₁₁H₁₃ClN₂O₃
MW: 256.686 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07629628
CAS Number: 133053-93-7