2,4-Dimethyl-5-(1H-pyrazol-3-yl)-1,3-thiazole
Supplier: Apollo Scientific
Warning
OR110950-1G OR110950-5G
APOSOR110950-1GEA 186 EUR
APOSOR110950-1G APOSOR110950-5G
2,4-Dimethyl-5-(1H-pyrazol-3-yl)-1,3-thiazole
2,4-Dimethyl-5-(1H-pyrazol-3-yl)-1,3-thiazole
Formula: C₈H₉N₃S
MW: 179.242 g/mol
Melting Pt: 161…164 °C
Storage Temperature: Ambient
MDL Number: MFCD01763614
CAS Number: 439107-00-3