Ethyl (4-hydroxy-3-nitrophenyl)acetate
Supplier: Apollo Scientific
Warning
OR110951-1G OR110951-5G OR110951-250MG
APOSOR110951-1GEA 58 EUR
APOSOR110951-1G APOSOR110951-5G APOSOR110951-250MG
Ethyl (4-hydroxy-3-nitrophenyl)acetate
Ethyl (4-hydroxy-3-nitrophenyl)acetate
Formula: C₁₀H₁₁NO₅
MW: 225.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00224669
CAS Number: 183380-81-6