Ethyl 4-chloro-1H-indole-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4-Chloro-2-indolecarboxylate
OR110957-1G OR110957-100MG OR110957-250MG
APOSOR110957-1GEA 69.6 EUR
APOSOR110957-1G APOSOR110957-100MG APOSOR110957-250MG
Ethyl 4-chloro-1H-indole-2-carboxylate
Ethyl 4-chloro-1H-indole-2-carboxylate
Formula: C₁₁H₁₀ClNO₂
MW: 223.656 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264531
CAS Number: 53590-46-8