4-Bromophenyl 3-bromopropyl ether
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-(3-bromopropoxy)benzene
OR110997-1G OR110997-10G OR110997-5G
APOSOR110997-1GEA 92.8 EUR
APOSOR110997-1G APOSOR110997-10G APOSOR110997-5G
4-Bromophenyl 3-bromopropyl ether
4-Bromophenyl 3-bromopropyl ether
Formula: C₉H₁₀Br₂O
MW: 293.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00596633
CAS Number: 7497-87-2