5-(2-Ethylpiperidin-1-yl)-1,3,4-thiadiazol-2-amine
Supplier: Apollo Scientific
OR111041-1G OR111041-5G
APOSOR111041-1GEA 0 EUR
APOSOR111041-1G APOSOR111041-5G
5-(2-Ethylpiperidin-1-yl)-1,3,4-thiadiazol-2-amine
5-(2-Ethylpiperidin-1-yl)-1,3,4-thiadiazol-2-amine
Formula: C₉H₁₆N₄S
MW: 212.315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28148244
CAS Number: 1502633-77-3