Methyl 3-hydroxyadamantane-1-carboxylate
Supplier: Apollo Scientific
Warning
OR111047-1G OR111047-5G
APOSOR111047-1GEA 58 EUR
APOSOR111047-1G APOSOR111047-5G
Methyl 3-hydroxyadamantane-1-carboxylate
Methyl 3-hydroxyadamantane-1-carboxylate
Formula: C₁₂H₁₈O₃
MW: 210.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01838249
CAS Number: 68435-07-4