N-(4-Chloro-2,5-dimethoxyphenyl)thiourea
Supplier: Apollo Scientific
OR111049-10G OR111049-1G OR111049-5G
APOSOR111049-10GEA 0 EUR
APOSOR111049-10G APOSOR111049-1G APOSOR111049-5G
N-(4-Chloro-2,5-dimethoxyphenyl)thiourea
N-(4-Chloro-2,5-dimethoxyphenyl)thiourea
Formula: C₉H₁₁ClN₂O₂S
MW: 246.714 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00175088
CAS Number: 72806-65-6