N-(4-Chloro-3-nitrophenyl)-1H-pyrazole-1-carboxamide
Supplier: Apollo Scientific
OR111082-1G OR111082-5G OR111082-10G
APOSOR111082-1GEA 0 EUR
APOSOR111082-1G APOSOR111082-5G APOSOR111082-10G
N-(4-Chloro-3-nitrophenyl)-1H-pyrazole-1-carboxamide
N-(4-Chloro-3-nitrophenyl)-1H-pyrazole-1-carboxamide
Formula: C₁₀H₇ClN₄O₃
MW: 266.641 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28347983
CAS Number: 1858249-82-7