N-(4-Chloro-3-nitrophenyl)-1H-imidazole-1-carboxamide
Supplier: Apollo Scientific
OR111083-1G OR111083-5G OR111083-10G
APOSOR111083-1GEA 0 EUR
APOSOR111083-1G APOSOR111083-5G APOSOR111083-10G
N-(4-Chloro-3-nitrophenyl)-1H-imidazole-1-carboxamide
N-(4-Chloro-3-nitrophenyl)-1H-imidazole-1-carboxamide
Formula: C₁₀H₇ClN₄O₃
MW: 266.641 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23543204
CAS Number: 1535430-29-5