1-(4-Chlorophenyl)-5-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylic acid
Supplier: Apollo Scientific
OR111090-1G OR111090-5G
APOSOR111090-1GEA 310 EUR
APOSOR111090-1G APOSOR111090-5G
1-(4-Chlorophenyl)-5-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylic acid
1-(4-Chlorophenyl)-5-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylic acid
Formula: C₁₆H₁₃ClN₂O₂
MW: 300.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18434019
CAS Number: 861151-23-7