1,3-Dimethyl-5-morpholino-1H-pyrazol-4-amine
Supplier: Apollo Scientific
Warning
OR111117-1G OR111117-5G
APOSOR111117-1GEA 232 EUR
APOSOR111117-1G APOSOR111117-5G
1,3-Dimethyl-5-morpholino-1H-pyrazol-4-amine
1,3-Dimethyl-5-morpholino-1H-pyrazol-4-amine
Formula: C₉H₁₆N₄O
MW: 196.25 g/mol
Melting Pt: 155.5…156.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04115380
CAS Number: 568577-87-7