1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carbaldehyde
Supplier: Apollo Scientific
Warning
OR111192-1G OR111192-5G
APOSOR111192-1GEA 447 EUR
APOSOR111192-1G APOSOR111192-5G
1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carbaldehyde
1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carbaldehyde
Formula: C₈H₈N₂OS
MW: 180.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03086154
CAS Number: 50880-19-8