4-Azepan-1-ylbenzonitrile

Supplier: Apollo Scientific

OR111240-10G OR111240-1G OR111240-5G
APOSOR111240-10GEA 0 EUR
APOSOR111240-10G APOSOR111240-1G APOSOR111240-5G
4-Azepan-1-ylbenzonitrile
4-Azepan-1-ylbenzonitrile
Formula: C₁₃H₁₆N₂
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11038470
CAS Number: 162377-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR