5-Methoxy-1-methyl-1H-indole
Supplier: Apollo Scientific
Warning
OR111390-10G OR111390-1G OR111390-5G
APOSOR111390-10GEA 464 EUR
APOSOR111390-10G APOSOR111390-1G APOSOR111390-5G
5-Methoxy-1-methyl-1H-indole
5-Methoxy-1-methyl-1H-indole
Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863250
CAS Number: 2521-13-3