2-(4-Nitrophenyl)-1,3-dioxolane
Supplier: Apollo Scientific
Warning
OR111541-1G OR111541-5G
APOSOR111541-1GEA 69.6 EUR
APOSOR111541-1G APOSOR111541-5G
2-(4-Nitrophenyl)-1,3-dioxolane
2-(4-Nitrophenyl)-1,3-dioxolane
Formula: C₉H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00550438
CAS Number: 2403-53-4