Isopropyl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate
Supplier: Apollo Scientific
Warning
Synonyms: S-(+)-Methoprene, Prop-2-yl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate
OR11200-1G OR11200-25G OR11200-5G
APOSOR11200-1GEA 37.7 EUR
APOSOR11200-1G APOSOR11200-25G APOSOR11200-5G
Isopropyl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate
Isopropyl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate
Formula: C₁₉H₃₄O₃
MW: 310.471 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00867612
CAS Number: 65733-16-6