1-Methyl-5-phenyl-1H-pyrazole-4-carboxylic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Carboxy-1-methyl-5-phenyl-1H-pyrazole
OR11281-1G OR11281-250MG
APOSOR11281-1GEA 435 EUR
APOSOR11281-1G APOSOR11281-250MG
1-Methyl-5-phenyl-1H-pyrazole-4-carboxylic acid 97%
1-Methyl-5-phenyl-1H-pyrazole-4-carboxylic acid
Formula: C₁₁H₁₀N₂O₂
MW: 202.209 g/mol
Melting Pt: 212…216 °C
Storage Temperature: Ambient
MDL Number: MFCD04038957
CAS Number: 105994-75-0