4-Chloro-6-(1H-pyrazol-1-yl)pyrimidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(6-Chloropyrimidin-4-yl)-1H-pyrazole
OR11468-1G OR11468-100MG
APOSOR11468-1GEA 174 EUR
APOSOR11468-1G APOSOR11468-100MG
4-Chloro-6-(1H-pyrazol-1-yl)pyrimidine 98%
4-Chloro-6-(1H-pyrazol-1-yl)pyrimidine
Formula: C₇H₅ClN₄
MW: 180.594 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00297274
CAS Number: 114833-95-3